Learn More
TARGETMOL CHEMICALS INC ETC-159 25MG
SDP
Catalog No. 502576822
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ETC-159 (ETC-1922159) is a potent orally available PORCN inhibitor. It inhibits (beta)-catenin reporter activity with an IC50 of 2.9 nM. purity: 96%
Catalog No. 50-257-6822 Supplier TARGETMOL CHEMICALS INC Supplier No. T404325MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ETC-159 (ETC-1922159) is a potent orally available PORCN inhibitor. It inhibits (beta)-catenin reporter activity with an IC50 of 2.9 nM. purity: 96%