Learn More
Selleck Chemical LLC ETC-159
SDP
Catalog No. 5000000497
Encompass_Preferred
ETC-159 (ETC-1922159) is an orally available potent porcupine inhibitor with an IC50 of 2 9 nM for inhibiting -catenin reporter activity in STF3A cells
Catalog No. 50-000-00497 Supplier Selleck Chemical LLC Supplier No. S661610MM/1ML
ETC-159 (ETC-1922159) is an orally available potent porcupine inhibitor with an IC50 of 2 9 nM for inhibiting -catenin reporter activity in STF3A cells