Learn More
Selleck Chemical LLC ETC-159
SDP
Catalog No. 5000002643
Encompass_Preferred
ETC-159 (ETC-1922159) is an orally available potent porcupine inhibitor with an IC50 of 2 9 nM for inhibiting -catenin reporter activity in STF3A cells
Catalog No. 50-000-02643 Supplier Selleck Chemical LLC Supplier No. S6616100MG
ETC-159 (ETC-1922159) is an orally available potent porcupine inhibitor with an IC50 of 2 9 nM for inhibiting -catenin reporter activity in STF3A cells