Learn More
Selleck Chemical LLC ETC-159 5mg
SDP
Supplier:  Selleck Chemical LLC S66165MG
Encompass_Preferred
ETC-159 (ETC-1922159) is an orally available, potent porcupine inhibitor with an IC50 of 2.9 nM for inhibiting ?-catenin reporter activity in STF3A cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3187