Learn More
Electron Microscopy Sciences Electrically/Thermally Conductive Adhesive ETC Bond 525 5 GR
SDP
Catalog No. 50997382
Encompass_Preferred
Electrically conductive adhesive, silver filled, one-part epoxy paste, good chemical resistance and mechanical strength to 340 deg F.
Catalog No. 50-997-382 Supplier Electron Microscopy Sciences Supplier No. 50380525
Electrically conductive adhesive, silver filled, one-part epoxy paste, good chemical resistance and mechanical strength to 340 deg F.