Learn More
Electron Microscopy Sciences Electrically/Thermally Conductive Adhesive ETC Bond 556 5 GR
SDP
Catalog No. 50997383
Encompass_Preferred
Electrically conductive adhesive, Silver filled, 1:1 Epoxy paste, Good corrosion resistance and mechanical strength to 340 deg F.
Catalog No. 50-997-383 Supplier Electron Microscopy Sciences Supplier No. 50380556
Electrically conductive adhesive, Silver filled, 1:1 Epoxy paste, Good corrosion resistance and mechanical strength to 340 deg F.