Learn More
Electron Microscopy Sciences Electrically/Thermally Conductive Adhesive ETC Bond 597a 5 GR
SDP
Catalog No. 50997385
Encompass_Preferred
Electrically and thermally conductive adhesive, Silver filled. One part system, Inorganic system for bonding applications up to 1200 deg F.
Catalog No. 50-997-385 Supplier Electron Microscopy Sciences Supplier No. 50380597
Electrically and thermally conductive adhesive, Silver filled. One part system, Inorganic system for bonding applications up to 1200 deg F.