Learn More
Transene Inc Chromium Etchant 1020, 1 Gallon
SDP
Catalog No. NC1412121
Encompass
Transene Chromium Etchants 1020 is a high purity ceric ammonium nitrate systems for precise, clean etching of chromium and chromium oxide films. The etchant is compatible with positive and negative photoresists. Chromium Etchant 1020 is filtered to remove all particulates above 0.2 microns.
This item is not returnable. View return policy
Catalog No. NC1412121 Supplier Transene Inc Supplier No. 0600021001GAL
Transene Chromium Etchants 1020 is a high purity ceric ammonium nitrate systems for precise, clean etching of chromium and chromium oxide films. The etchant is compatible with positive and negative photoresists. Chromium Etchant 1020 is filtered to remove all particulates above 0.2 microns.