Learn More
Transene Inc Nickel Etchant TFB | 1 Gallon
SDP
Catalog No. NC1412119
Encompass_Preferred
Standard nickel etchant for evaporated nickel films.
Catalog No. NC1412119 Supplier Transene Inc Supplier No. 0600016000GAL
Standard nickel etchant for evaporated nickel films.