Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Etchant (Ferric Chloride/Hydrochloric Acid), Reagents

Catalog No. 01335374
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-374 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-374 Supplier Reagents Holdings Llc Supplier No. CE2325001A
Add to Cart
Add to Cart

Etchant (Ferric Chloride/Hydrochloric Acid), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Color Orange
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.0717
pH <1
Quantity 1 L
Sterility Non-sterile
Physical Form Liquid
Chemical Name or Material Etchant (Ferric Chloride/Hydrochloric Acid)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.