Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Etching Solution for Stainless Steel, Reagents

Catalog No. 01335371
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-371 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-371 Supplier Reagents Holdings Llc Supplier No. CE237500500A
Add to Cart
Add to Cart

Etching Solution for Stainless Steel, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Etching Solution for Stainless Steel
Color Brown/Yellow
CAS 7647-01-0, 10025-77-1
Boiling Point 100°C
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.3258
pH <1
Quantity 500 mL
Sterility Non-sterile
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.