Learn More
Waters Corp Service Kit, Rebuild Kit, MS, Annual
Catalog No. 502392784 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
  • Service kit for use in maintaining an ETD Source
  • Kit includes ETD Glow Discharge Pin
  • Kit includes o-ring
  • Kit includes loop assembly
  • Type: Rebuild kit
  • System type: MS
  • Recommended for annual replacement
Catalog No. 50-239-2784 Supplier Waters Corp Supplier No. 700005276
  • Service kit for use in maintaining an ETD Source
  • Kit includes ETD Glow Discharge Pin
  • Kit includes o-ring
  • Kit includes loop assembly
  • Type: Rebuild kit
  • System type: MS
  • Recommended for annual replacement