Learn More
Med Vet International ETDA K3 Blood Collection Tube, Purple Cap, 3ml, J1560, Each
Catalog No. 5000583626
Encompass_Preferred
JorVet offers a wide range of blood collection tubes in various sizes and formats. All tubes adhere to the high standards of human blood collection tubes. Plastic versus Glass Human blood collection tubes have generally switched over to PET plastic tubes versus breakable glass. JorVet also uses the latest standardized color plastic caps for consistency. All tubes are in storage tray of 100 tubes K3 EDTA Tubes are clot prevention in tubes often used for CBC's ETDA K3 Blood Collection Tube - 3ml.
Catalog No. 50-005-83626 Supplier Med Vet International Supplier No. JOR1560
JorVet offers a wide range of blood collection tubes in various sizes and formats. All tubes adhere to the high standards of human blood collection tubes. Plastic versus Glass Human blood collection tubes have generally switched over to PET plastic tubes versus breakable glass. JorVet also uses the latest standardized color plastic caps for consistency. All tubes are in storage tray of 100 tubes K3 EDTA Tubes are clot prevention in tubes often used for CBC's ETDA K3 Blood Collection Tube - 3ml.