Learn More
Medchemexpress LLC Etelcalcetide (hydrochloride) | 1334237-71-6 | 87645 | 99.3% | 1353.33 | 25 MG
SDP
Catalog No. 5000426692
Encompass_Preferred
Etelcalcetide hydrochloride is a synthetic calcimimetic that acts as an activator of the calcium sensing receptor (CaSR). It is effective in lowering parathyroid hormone (PTH) concentrations in dialysis patients with secondary hyperparathyroidism undergoing hemodialysis, showing promise for research in secondary hyperparathyroidism and chronic kidney disease.
  • Activates the calcium sensing receptor (CaSR)
  • Lowers parathyroid hormone (PTH) concentrations
  • Effective for secondary hyperparathyroidism in dialysis patients
  • Useful for research in secondary hyperparathyroidism and chronic kidney disease
  • Reduces parathyroid hormone (PTH) levels in human parathyroid cells in vitro
  • Significantly reduces plasma parathyroid hormone (PTH) concentrations in vivo
Catalog No. 50-004-26692 Supplier Medchemexpress LLC Supplier No. 25MGHYP1955A
Etelcalcetide hydrochloride is a synthetic calcimimetic that acts as an activator of the calcium sensing receptor (CaSR). It is effective in lowering parathyroid hormone (PTH) concentrations in dialysis patients with secondary hyperparathyroidism undergoing hemodialysis, showing promise for research in secondary hyperparathyroidism and chronic kidney disease.
  • Activates the calcium sensing receptor (CaSR)
  • Lowers parathyroid hormone (PTH) concentrations
  • Effective for secondary hyperparathyroidism in dialysis patients
  • Useful for research in secondary hyperparathyroidism and chronic kidney disease
  • Reduces parathyroid hormone (PTH) levels in human parathyroid cells in vitro
  • Significantly reduces plasma parathyroid hormone (PTH) concentrations in vivo