Learn More
Medchemexpress LLC Etelcalcetide hydrochloride | 1334237-71-6 | 99.80% | 1353.33 | 50 MG
SDP
Catalog No. 5000422883
Encompass_Preferred
Etelcalcetide hydrochloride is a synthetic calcimimetic that acts as an activator of the calcium sensing receptor (CaSR), effective in lowering parathyroid hormone (PTH) concentrations in dialysis patients with secondary hyperparathyroidism.
  • Reduces parathyroid hormone (PTH) levels in human parathyroid cells in a dose-dependent manner.
  • Significantly reduces plasma PTH concentrations in C57BL7/6J male mice.
  • Decreases renal expression of Cyp24a1.
  • Promising for research in secondary hyperparathyroidism and chronic kidney disease.
Catalog No. 50-004-22883 Supplier Medchemexpress LLC Supplier No. HYP1955A50MG
Etelcalcetide hydrochloride is a synthetic calcimimetic that acts as an activator of the calcium sensing receptor (CaSR), effective in lowering parathyroid hormone (PTH) concentrations in dialysis patients with secondary hyperparathyroidism.
  • Reduces parathyroid hormone (PTH) levels in human parathyroid cells in a dose-dependent manner.
  • Significantly reduces plasma PTH concentrations in C57BL7/6J male mice.
  • Decreases renal expression of Cyp24a1.
  • Promising for research in secondary hyperparathyroidism and chronic kidney disease.