Learn More
Cayman Chemical EtelcalcetIdehydrochLRIde 10mg
SDP
Catalog No. 502937641
Encompass_Preferred
A peptide agonist of CaSR; selective for CaSR over a panel of 33 receptors and ion channels, as well as the norepinephrine transporter at 10 µM; increases intracellular calcium levels in HEK293T cells expressing the human CaSR receptor (EC50 = 0.53 µM); inhibits parathyroid secretion from primary rat parathyroid cells (EC50 = 0.36 µM in the presence of calcium); decreases parathyroid hormone and calcium in plasma and serum, respectively, in a model of chronic renal insufficiency with secondary hyperthyroidism in nephrectomized rats fed high-phosphorus diet at 0.3, 1, and 3 mg/kg
Catalog No. 50-293-7641 Supplier Cayman Chemical Supplier No. 2690110 MG
A peptide agonist of CaSR; selective for CaSR over a panel of 33 receptors and ion channels, as well as the norepinephrine transporter at 10 µM; increases intracellular calcium levels in HEK293T cells expressing the human CaSR receptor (EC50 = 0.53 µM); inhibits parathyroid secretion from primary rat parathyroid cells (EC50 = 0.36 µM in the presence of calcium); decreases parathyroid hormone and calcium in plasma and serum, respectively, in a model of chronic renal insufficiency with secondary hyperthyroidism in nephrectomized rats fed high-phosphorus diet at 0.3, 1, and 3 mg/kg