Learn More
Medchemexpress LLC Eteplirsen | 1173755-55-9 | 98.5% | 10306.00 | 50 MG
SDP
Catalog No. 5000344532
Encompass_Preferred
Eteplirsen (AVI 4658) is a synthetic antisense oligonucleotide for Duchenne muscular dystrophy research. It promotes dystrophin production by restoring the translational reading frame of Duchenne muscular dystrophy (DMD) through specific skipping of exon 51 in defective gene variants.
  • Used for Duchenne muscular dystrophy research
  • Promotes dystrophin production
  • Restores translational reading frame of DMD
  • Achieves specific skipping of exon 51 in defective gene variants
Catalog No. 50-003-44532 Supplier Medchemexpress LLC Supplier No. HY10875310MG
Eteplirsen (AVI 4658) is a synthetic antisense oligonucleotide for Duchenne muscular dystrophy research. It promotes dystrophin production by restoring the translational reading frame of Duchenne muscular dystrophy (DMD) through specific skipping of exon 51 in defective gene variants.
  • Used for Duchenne muscular dystrophy research
  • Promotes dystrophin production
  • Restores translational reading frame of DMD
  • Achieves specific skipping of exon 51 in defective gene variants