Learn More
Medchemexpress LLC Eteplirsen sodium | 91.2% | 10306 | 1 MG
SDP
Catalog No. 5000344403
Encompass_Preferred
Eteplirsen (AVI 4658) sodium is a synthetic antisense oligonucleotide that induces dystrophin production. It promotes exon 51 skipping in Duchenne muscular dystrophy patients and is used in Duchenne muscular dystrophy research.
  • Induces dystrophin production.
  • Promotes exon 51 skipping.
  • Research use.
  • High purity.
Catalog No. 50-003-44403 Supplier Medchemexpress LLC Supplier No. HY108753A1MG
Eteplirsen (AVI 4658) sodium is a synthetic antisense oligonucleotide that induces dystrophin production. It promotes exon 51 skipping in Duchenne muscular dystrophy patients and is used in Duchenne muscular dystrophy research.
  • Induces dystrophin production.
  • Promotes exon 51 skipping.
  • Research use.
  • High purity.