Learn More
Aviva Systems Biology Corp ETFB Antibody - C-terminal region (ARP54442_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54442_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ETFB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ETFB. Alternate names: FP585, MADDApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-61207