Learn More
Analytical Sales & Services ETFE Ferrule for 1/16" OD Tubing
Catalog No. 5000804671
Encompass
ETFE Flangeless Ferrule for 1/16" OD Tubing Red
Qty/Pkg: EACH
Catalog No. 50-008-04671 Supplier Analytical Sales & Services Supplier No. 59200R
ETFE Flangeless Ferrule for 1/16" OD Tubing Red
Qty/Pkg: EACH