Learn More
Analytical Sales & Services ETFE Ferrule with SS Locking Ring
Catalog No. 5000804666
Encompass
Yellow ETFE Ferrule w/ Stainless Steel Locking Ring, 1/4-28 Flat-Bottom Port
Catalog No. 50-008-04666 Supplier Analytical Sales & Services Supplier No. 57250X
Yellow ETFE Ferrule w/ Stainless Steel Locking Ring, 1/4-28 Flat-Bottom Port