Learn More
Waters Corp Waters Corporation Tubing, ETFE, 1/16 in. OD x 0.010 in. ID, 204 mm long
Catalog No. 50466648 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This ETFE tubing is designed for various laboratory applications. It features precise dimensions of 1/16 in. outer diameter and 0.010 in. inner diameter, with a length of 204 mm.
  • ETFE material ensures chemical resistance and durability.
  • Outer diameter of 1/16 in. and inner diameter of 0.010 in.
  • Length of 204 mm for versatile applications.
  • Provided as a spare part kit for convenience.
  • Includes one unit per package.
  • Designed for replacement as required to maintain system performance.
Catalog No. 50-466-648 Supplier Waters Corp Supplier No. 700005363
This ETFE tubing is designed for various laboratory applications. It features precise dimensions of 1/16 in. outer diameter and 0.010 in. inner diameter, with a length of 204 mm.
  • ETFE material ensures chemical resistance and durability.
  • Outer diameter of 1/16 in. and inner diameter of 0.010 in.
  • Length of 204 mm for versatile applications.
  • Provided as a spare part kit for convenience.
  • Includes one unit per package.
  • Designed for replacement as required to maintain system performance.