Learn More
Cliawaived Inc Etg (Ethyl Glucuronide) Rapid Tests
SDP
Supplier:  Cliawaived Inc AVETG500
Encompass_Preferred
(25 Tests per Box)New Etg rapid urine testing strips. Detects prior alcohol use up to 80 hours. Read results at 5 minutes. 500 ng/mL cut-off. High quality results and comparable to lab tests.Note: This test is for forensic use only. This test is not FDA Cleared. This test is not CLIA waived. This test may not be used for clinical diagnosis or treatment.
Catalog No. NC1859177