Learn More
Medchemexpress LLC Eth 157 | 61595-77-5 | 556.65 | 25 MG
SDP
Catalog No. 5000347919
Encompass_Preferred
ETH 157 is a Na+ ionophore that induces a Na+/Ca2+ selectivity in membranes. This product is for research use only.
  • Acts as a Na+ ionophore
  • Induces Na+/Ca2+ selectivity in membranes
  • Targets the sodium channel
  • Pathway: membrane transporter/ion channel
  • Store powder at -20°C for 3 years, or 4°C for 2 years
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month
Catalog No. 50-003-47919 Supplier Medchemexpress LLC Supplier No. 25MGHY137172
ETH 157 is a Na+ ionophore that induces a Na+/Ca2+ selectivity in membranes. This product is for research use only.
  • Acts as a Na+ ionophore
  • Induces Na+/Ca2+ selectivity in membranes
  • Targets the sodium channel
  • Pathway: membrane transporter/ion channel
  • Store powder at -20°C for 3 years, or 4°C for 2 years
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month