Learn More
Medchemexpress LLC ETH 157 | 61595-77-5 | 556.65 | 50 MG
SDP
Catalog No. 5000351095
Encompass_Preferred
ETH 157 is a Na+ ionophore that induces a Na+/Ca2+ selectivity in membranes. It is intended for research use only.
  • Na+ ionophore
  • Induces Na+/Ca2+ selectivity in membranes
Catalog No. 50-003-51095 Supplier Medchemexpress LLC Supplier No. HY13717250MG
ETH 157 is a Na+ ionophore that induces a Na+/Ca2+ selectivity in membranes. It is intended for research use only.
  • Na+ ionophore
  • Induces Na+/Ca2+ selectivity in membranes