Learn More
Cayman Chemical ETH-LAdolutIon 100ug
SDP
Catalog No. 502944742
Encompass_Preferred
An analytical reference standard categorized as a lysergamide; substitutes for LSD (D-tartrate) in a two-lever drug discrimination test in rats; intended for research and forensic applications
Catalog No. 50-294-4742 Supplier Cayman Chemical Supplier No. 41271100 UG
An analytical reference standard categorized as a lysergamide; substitutes for LSD (D-tartrate) in a two-lever drug discrimination test in rats; intended for research and forensic applications