Learn More
Cayman Chemical Ethacridine (lactate), Purity: ≥98%, CAS: 1837-57-6, 5 g
SDP
Catalog No. 502944862
Encompass_Preferred
An inducer of uterine contraction; induces contractions in primary mouse myometrium smooth muscle cells when co-cultured with primary degranulated mast cell and isolated and washed mouse uterine horns at 10 µM; preincubation of viral particles reduces SARS-CoV-2 viral titers in a plaque assay, as well as decreases the levels of intracellular and extracellular viral RNA and the intracellular levels of the N protein and S protein, in Vero E6 cells at 5 µM; decreases colony formation by, and induces apoptosis in, SW1736 and FTC-133 cancer cells at 10 µM; reduces the levels of mRNA encoding TTF-1, Pax8, and NIS in Nthy-ori 3-1 thyroid follicular epithelial cells at 10 µM
Catalog No. 50-294-4862 Supplier Cayman Chemical Supplier No. 402825 G
An inducer of uterine contraction; induces contractions in primary mouse myometrium smooth muscle cells when co-cultured with primary degranulated mast cell and isolated and washed mouse uterine horns at 10 µM; preincubation of viral particles reduces SARS-CoV-2 viral titers in a plaque assay, as well as decreases the levels of intracellular and extracellular viral RNA and the intracellular levels of the N protein and S protein, in Vero E6 cells at 5 µM; decreases colony formation by, and induces apoptosis in, SW1736 and FTC-133 cancer cells at 10 µM; reduces the levels of mRNA encoding TTF-1, Pax8, and NIS in Nthy-ori 3-1 thyroid follicular epithelial cells at 10 µM