Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethacrynic acid | 58-54-8 | MFCD00056693 | 98.8% | 303.14 g/mol | C13H12Cl2O4 | 10 MG
SDP

Catalog No. 5000297270
Encompass_Preferred

Ethacrynic acid analytical standard supplied for research and analytical use. The compound is a loop diuretic and bioactive small molecule reported to modulate glutathione S-transferases and NF-κB signaling, affect calcium channels, and show anti-inflammatory activity; it is provided as a purified solid standard for biochemical and pharmacological studies.

  • Intended for research and analytical applications.
  • Purity approximately 98.8%.
  • Molecular weight 303.14 g/mol.
  • CAS number 58-54-8 for unambiguous identification.
  • Available in small package sizes, including 10 mg.
  • Supplied as a solid powder suitable for analytical preparation.

Catalog No. 50-002-97270 Supplier Medchemexpress LLC Supplier No. HYB1640R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethacrynic acid analytical standard supplied for research and analytical use. The compound is a loop diuretic and bioactive small molecule reported to modulate glutathione S-transferases and NF-κB signaling, affect calcium channels, and show anti-inflammatory activity; it is provided as a purified solid standard for biochemical and pharmacological studies.

  • Intended for research and analytical applications.
  • Purity approximately 98.8%.
  • Molecular weight 303.14 g/mol.
  • CAS number 58-54-8 for unambiguous identification.
  • Available in small package sizes, including 10 mg.
  • Supplied as a solid powder suitable for analytical preparation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.