Learn More
Enzo Life Sciences Ethacrynic acid, (1g), CAS Number: 58-54-8
SDP
Supplier:  Enzo Life Sciences BMLEI1280001
Encompass_Preferred
GST inhibitor. Inhibits LTC4 biosynthesis by inhibition of glutathione S-transferase (GST). Inhibits 15-hydroxyprostaglandin dehydrogenase. Alternative Name: 2,3-Dichloro-4-(2-methylene-1-oxobutyl)phenoxyacetic acid. MW: 303.1. CAS Number: 58-54-8. Purity: ≥98%. Appearance: Off-white to white. Solubility: Soluble in 100% ethanol (35mg/ml) or DMSO (35mg/ml). Long Term Storage: Ambient
Catalog No. NC0267715