Learn More
Selleck Chemical LLC Ethambutol 2HCl S4004-50mg
SDP
Catalog No. 503108833
Encompass_Preferred
Ethambutol 2HCl (CL40881) is a bacteriostatic antimycobacterial agent which obstructs the formation of cell wall by inhibiting arabinosyl transferases
Catalog No. 50-310-8833 Supplier Selleck Chemical LLC Supplier No. S400450MG
Ethambutol 2HCl (CL40881) is a bacteriostatic antimycobacterial agent which obstructs the formation of cell wall by inhibiting arabinosyl transferases