Learn More
Medchemexpress LLC Ethambutol-d10 | 1129526-24-4 | 99.0% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0535S11MG
Encompass_Preferred
Ethambutol-d10 is a deuterium-labeled version of Ethambutol, a bacteriostatic antimycobacterial agent that inhibits arabinosyl transferases, thereby obstructing the formation of the cell wall. Intended for research use only and not sold to patients, stable heavy isotopes like deuterium are incorporated into drug molecules as tracers for quantitation during drug development and have the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-07619