Learn More
Medchemexpress LLC Ethambutol-d4 | 1129526-19-7 | 98.1% | 1 MG
SDP
Catalog No. 5000408845
Encompass_Preferred
Ethambutol-d4 is a deuterium-labeled version of Ethambutol, a bacteriostatic antimycobacterial agent. It functions by inhibiting arabinosyl transferases, thereby obstructing the formation of bacterial cell walls. This compound can be utilized as a tracer and as an internal standard for quantitative analysis in various methods.
  • Deuterium-labeled ethambutol
  • Bacteriostatic antimycobacterial agent
  • Inhibits arabinosyl transferases
  • Obstructs bacterial cell wall formation
  • Suitable for use as a tracer
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-08845 Supplier Medchemexpress LLC Supplier No. HYB0535S1MG
Ethambutol-d4 is a deuterium-labeled version of Ethambutol, a bacteriostatic antimycobacterial agent. It functions by inhibiting arabinosyl transferases, thereby obstructing the formation of bacterial cell walls. This compound can be utilized as a tracer and as an internal standard for quantitative analysis in various methods.
  • Deuterium-labeled ethambutol
  • Bacteriostatic antimycobacterial agent
  • Inhibits arabinosyl transferases
  • Obstructs bacterial cell wall formation
  • Suitable for use as a tracer
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS