Learn More
Medchemexpress LLC Ethambutol-d4 | 1129526-19-7 | 98.12% | 5 MG
SDP
Catalog No. 5000405851
Encompass_Preferred
Ethambutol-d4 is the deuterium-labeled version of Ethambutol, a bacteriostatic antimycobacterial agent. It obstructs the formation of the cell wall by inhibiting arabinosyl transferases. This compound is intended for research use only and can be utilized as a tracer or an internal standard for quantitative analysis.
  • Used as a tracer in drug development
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium-labeled compound for studying pharmacokinetic and metabolic profiles
Catalog No. 50-004-05851 Supplier Medchemexpress LLC Supplier No. HYB0535S5MG
Ethambutol-d4 is the deuterium-labeled version of Ethambutol, a bacteriostatic antimycobacterial agent. It obstructs the formation of the cell wall by inhibiting arabinosyl transferases. This compound is intended for research use only and can be utilized as a tracer or an internal standard for quantitative analysis.
  • Used as a tracer in drug development
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium-labeled compound for studying pharmacokinetic and metabolic profiles