Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethambutol DiHCl, G-Biosciences
SCI_ED

Catalog No. S139800
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139800 100 g
S139732 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S139800 Supplier G-Biosciences Supplier No. RC1017
Add to Cart
Add to Cart

Ethambutol DiHCl is a hydrochloride salt form of an ethylenediamine derivative that is used as an anti-mycobiotic.

It has been used for studying multidrug resistance in Mycobacterium tuberculosis and to investigate effective antibiotic combinations for ethambutol DiHCl.

Chemical Identifiers

Molecular Weight (g/mol) 277.23 g/mol

Specifications

Boiling Point 345°C
Quantity 100 g
Percent Purity 98%
Grade Laboratory
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.