Learn More
Medchemexpress LLC Ethambutol dihydrochloride | 1070-11-7 | 99.9% | 5 G
SDP
Catalog No. 5000407618
Encompass_Preferred
Ethambutol dihydrochloride is an antimycobacterial compound that prevents cell wall formation by inhibiting arabinosyltransferase activity. It can be used to establish animal models of hyperuricemia and optic neuropathy.
  • Antimycobacterial compound
  • Prevents cell wall formation
  • Inhibits arabinosyltransferase activity
  • Used to establish animal models of hyperuricemia
  • Used to establish animal models of optic neuropathy
Catalog No. 50-004-07618 Supplier Medchemexpress LLC Supplier No. HYB0535A5G
Ethambutol dihydrochloride is an antimycobacterial compound that prevents cell wall formation by inhibiting arabinosyltransferase activity. It can be used to establish animal models of hyperuricemia and optic neuropathy.
  • Antimycobacterial compound
  • Prevents cell wall formation
  • Inhibits arabinosyltransferase activity
  • Used to establish animal models of hyperuricemia
  • Used to establish animal models of optic neuropathy