Learn More
Medchemexpress LLC Ethambutol dihydrochloride | 1070-11-7 | 99.9% | 50 MG
SDP
Catalog No. 5000406009
Encompass_Preferred
Ethambutol dihydrochloride is an antimycobacterial compound that prevents cell wall formation by inhibiting arabinosyltransferase activity. It can be used to establish animal models of hyperuricemia and optic neuropathy. This product is for research use only and not sold to patients.
  • Antimycobacterial compound
  • Prevents cell wall formation by inhibiting arabinosyltransferase activity
  • Used to establish animal models of hyperuricemia
  • Used to establish animal models of optic neuropathy
Catalog No. 50-004-06009 Supplier Medchemexpress LLC Supplier No. HYB0535AR50MG
Ethambutol dihydrochloride is an antimycobacterial compound that prevents cell wall formation by inhibiting arabinosyltransferase activity. It can be used to establish animal models of hyperuricemia and optic neuropathy. This product is for research use only and not sold to patients.
  • Antimycobacterial compound
  • Prevents cell wall formation by inhibiting arabinosyltransferase activity
  • Used to establish animal models of hyperuricemia
  • Used to establish animal models of optic neuropathy