Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethambutol, G-Biosciences
SCI_ED

Catalog No. S139819
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139819 25 g
S139731 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S139819 Supplier G-Biosciences Supplier No. RC1014
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethambutol is an antitubercular antibiotic effective against Mycobacterium species, especially Mycobacterium tuberculosis.

It has been used to study multidrug-resistant tuberculosis and in combination with other drugs to decrease the risk of developing additional multidrug resistant species.

Chemical Identifiers

Molecular Weight (g/mol) 204.31

Specifications

Physical Form Powder
Quantity 25 g
Product Line Gibco
Boiling Point 345.3°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.