Learn More
Apexbio Technology LLC Ethambutol HCl 1070-11-7 1g
SDP
Catalog No. 5000093860
Encompass_Preferred
Ethambutol HCl (CAS 1070-11-7) is a small-molecule inhibitor targeting mycobacterial arabinosyl transferase enzymes It is designed to inhibit this enzyme thereby disrupting mycobacterial cell wall biosynthesis Ethambutol HCl exerts its biological activity primarily through inhibition of arabinosyl transferase impairing cell wall integrity In in vitro studies Ethambutol HCl demonstrates bacteriostatic activity against actively replicating mycobacteria with an IC50 value in the micromolar concentration range Based on these pharmacological properties Ethambutol HCl holds research potential in tuberculosis studies including the evaluation of antimycobacterial efficacy and investigation of resistance mechanisms
Catalog No. 50-000-93860 Supplier Apexbio Technology LLC Supplier No. A84271000
Ethambutol HCl (CAS 1070-11-7) is a small-molecule inhibitor targeting mycobacterial arabinosyl transferase enzymes It is designed to inhibit this enzyme thereby disrupting mycobacterial cell wall biosynthesis Ethambutol HCl exerts its biological activity primarily through inhibition of arabinosyl transferase impairing cell wall integrity In in vitro studies Ethambutol HCl demonstrates bacteriostatic activity against actively replicating mycobacteria with an IC50 value in the micromolar concentration range Based on these pharmacological properties Ethambutol HCl holds research potential in tuberculosis studies including the evaluation of antimycobacterial efficacy and investigation of resistance mechanisms