Learn More
Sigma Aldrich Fine Chemicals Biosciences Ethambutol Related Compound A United States Pharmacopeia (USP) Reference Standard | 10054-06-5 | 15MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 125701815MG
Encompass
Ethambutol Related Compound A United States Pharmacopeia (USP) Reference Standard | Mol Wt: 204.31 | 10054-06-5 | 15MG
Catalog No. 50-186-1655