Learn More
Cayman Chemical Ethambutol (hydrochloride), Purity: ≥98%, CAS: 1070-11-7, 1 g
SDP
Catalog No. 502994969
Encompass_Preferred
An antimycobacterial compound; inhibits the growth of M. tuberculosis in vitro at a concentration of 10 μg/ml; protective against M. smegmatis infections in rhesus monkeys (50, 30, and 0% fatalities at doses of 12.5, 25, and 50 mg/kg, respectively); clears M. smegmatis pulmonary infiltrates at a dose of 100 mg/kg
Catalog No. 50-299-4969 Supplier Cayman Chemical Supplier No. 237131 G
An antimycobacterial compound; inhibits the growth of M. tuberculosis in vitro at a concentration of 10 μg/ml; protective against M. smegmatis infections in rhesus monkeys (50, 30, and 0% fatalities at doses of 12.5, 25, and 50 mg/kg, respectively); clears M. smegmatis pulmonary infiltrates at a dose of 100 mg/kg