Learn More
eMolecules​ Ethanamine, 2-[2-[2-[(6-chlorohexyl)oxy]ethoxy]ethoxy]-, hydrochloride (1:1) | Combi-Blocks, Inc. | 1261238-19-0 | | 304.250 | C12H27Cl2NO3 | 95.000 | Cl.NCCOCCOCCOCCCCCCCl | 100mg | 642124027
Supplier:  eMolecules​ 642124027
Encompass_Preferred
Ethanamine, 2-[2-[2-[(6-chlorohexyl)oxy]ethoxy]ethoxy]-, hydrochloride (1:1) | Combi-Blocks, Inc. | 1261238-19-0 | | 304.250 | C12H27Cl2NO3 | 95.000 | Cl.NCCOCCOCCOCCCCCCCl | 100mg | 642124027
Catalog No. NC2310717