Learn More
Medchemexpress LLC Ethanamine, 2-[2-[2-[(6-chlorohexyl)oxy]ethoxy]ethoxy]- | 1261350-60-0 | 95.0% | 267.79 | 25 MG
SDP
Catalog No. 5000352443
Encompass_Preferred
NH2-PEG3-C6-Cl is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker; one targets an E3 ubiquitin ligase, and the other targets a specific protein. This mechanism leverages the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins
Catalog No. 50-003-52443 Supplier Medchemexpress LLC Supplier No. 25MGHY138469
NH2-PEG3-C6-Cl is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker; one targets an E3 ubiquitin ligase, and the other targets a specific protein. This mechanism leverages the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins