Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethanamine, 2-[2-(2-propyn-1-yloxy)ethoxy]- | 944561-44-8 | 99.6% | 143.18 | C7H13NO2 | 1 G
SDP

Catalog No. 5000203119
Encompass_Preferred

Propargyl-PEG2-amine is a short PEG2-based linker containing a terminal propargyl (alkyne) group and a primary amine for bioconjugation. It is supplied as a colorless to light yellow liquid and is used as an ADC linker, PROTAC linker, and general crosslinking building block for click-type chemistries.

  • Contains a terminal propargyl functional group for click reactions.
  • Provides a short, flexible PEG2 spacer between functional groups.
  • Features a primary amine for coupling to carboxyl or activated esters.
  • Supplied as a colorless to light yellow liquid for easy handling.
  • Molecular weight 143.18 and CAS number 944561-44-8 for unambiguous identification.

Catalog No. 50-002-03119 Supplier Medchemexpress LLC Supplier No. HYW0516341G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-PEG2-amine is a short PEG2-based linker containing a terminal propargyl (alkyne) group and a primary amine for bioconjugation. It is supplied as a colorless to light yellow liquid and is used as an ADC linker, PROTAC linker, and general crosslinking building block for click-type chemistries.

  • Contains a terminal propargyl functional group for click reactions.
  • Provides a short, flexible PEG2 spacer between functional groups.
  • Features a primary amine for coupling to carboxyl or activated esters.
  • Supplied as a colorless to light yellow liquid for easy handling.
  • Molecular weight 143.18 and CAS number 944561-44-8 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.