Learn More
Medchemexpress LLC Ethanamine, 2-[2-(2-propyn-1-yloxy)ethoxy]- | 944561-44-8 | MFCD19160595 | 99.6% | 143.18 g/mol | C7H13NO2 | 100 G
SDP
Catalog No. 5000202861
Encompass_Preferred
Propargyl-PEG2-amine is a PEG-based bifunctional linker that contains a terminal propargyl (alkyne) group and a primary amine, designed for conjugation chemistry and synthetic applications such as antibody-drug conjugate (ADC) formation, PROTAC assembly, and click chemistry.
  • PEG-2 spacer with terminal propargyl group
  • Primary amine functional group for conjugation
  • High purity suitable for synthesis (99.6%)
  • Colorless to light yellow liquid; density 0.992 ± 0.06 g/cm3
  • Compatible with copper-catalyzed azide-alkyne cycloaddition (CuAAC)
Catalog No. 50-002-02861 Supplier Medchemexpress LLC Supplier No. HYW051634100G
Propargyl-PEG2-amine is a PEG-based bifunctional linker that contains a terminal propargyl (alkyne) group and a primary amine, designed for conjugation chemistry and synthetic applications such as antibody-drug conjugate (ADC) formation, PROTAC assembly, and click chemistry.
  • PEG-2 spacer with terminal propargyl group
  • Primary amine functional group for conjugation
  • High purity suitable for synthesis (99.6%)
  • Colorless to light yellow liquid; density 0.992 ± 0.06 g/cm3
  • Compatible with copper-catalyzed azide-alkyne cycloaddition (CuAAC)