Learn More
Medchemexpress LLC Ethanamine, 2-[2-(2-propyn-1-yloxy)ethoxy]- | 944561-44-8 | MFCD19160595 | 99.6% | 143.18 g/mol | C7H13NO2 | 250 MG
SDP
Catalog No. 5000202866
Encompass_Preferred
Propargyl-PEG2-amine is a PEG-based propargyl-functionalized amine used as a short spacer linker for bioconjugation. It provides a terminal alkyne for click chemistry and a primary amine for conjugation, making it suitable for linker synthesis and medicinal chemistry research.
  • Short PEG2 spacer with terminal alkyne for click chemistry.
  • Primary amine for facile conjugation to activated esters and similar chemistries.
  • High purity suitable for research-grade applications.
  • Compact 250 MG packaging for laboratory-scale experiments.
  • Soluble in common organic solvents used in synthesis and bioconjugation.
Catalog No. 50-002-02866 Supplier Medchemexpress LLC Supplier No. HYW051634250MG
Propargyl-PEG2-amine is a PEG-based propargyl-functionalized amine used as a short spacer linker for bioconjugation. It provides a terminal alkyne for click chemistry and a primary amine for conjugation, making it suitable for linker synthesis and medicinal chemistry research.
  • Short PEG2 spacer with terminal alkyne for click chemistry.
  • Primary amine for facile conjugation to activated esters and similar chemistries.
  • High purity suitable for research-grade applications.
  • Compact 250 MG packaging for laboratory-scale experiments.
  • Soluble in common organic solvents used in synthesis and bioconjugation.