Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethanamine, 2-(2-methoxyethoxy)- | 31576-51-9 | 99.9% | 119.16 | C5H13NO2 | 500 MG
SDP

Catalog No. 5000202840
Encompass_Preferred

m-PEG2-Amine is a methoxy-terminated polyethylene glycol (PEG) linker that provides a short PEG2 spacer with a terminal primary amine for conjugation. It is used as a cleavable linker in antibody-drug conjugate (ADC) synthesis and as a PEG-based linker in proteolysis-targeting chimera (PROTAC) construction.

  • Short PEG spacer with terminal primary amine for conjugation chemistry.
  • High purity (99.9% by GC) suitable for research applications.
  • Molecular weight 119.16 and formula C5H13NO2.
  • Available in small lab quantities, e.g., 500 MG.
  • Recommended storage: 4°C, protect from light; in solvent store at -80°C for long-term.

Catalog No. 50-002-02840 Supplier Medchemexpress LLC Supplier No. HYW008429500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG2-Amine is a methoxy-terminated polyethylene glycol (PEG) linker that provides a short PEG2 spacer with a terminal primary amine for conjugation. It is used as a cleavable linker in antibody-drug conjugate (ADC) synthesis and as a PEG-based linker in proteolysis-targeting chimera (PROTAC) construction.

  • Short PEG spacer with terminal primary amine for conjugation chemistry.
  • High purity (99.9% by GC) suitable for research applications.
  • Molecular weight 119.16 and formula C5H13NO2.
  • Available in small lab quantities, e.g., 500 MG.
  • Recommended storage: 4°C, protect from light; in solvent store at -80°C for long-term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.