Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethanamine, 2-(phenylmethoxy)- | 38336-04-8 | MFCD00235180 | 99.8% | 151.21 g/mol | C9H13NO | 50 G
SDP

Catalog No. 5000301647
Encompass_Preferred

2-(Benzyloxy)-1-ethanamine is a small-molecule biochemical reagent used as an organic building block and reagent in life-science and synthetic chemistry research. It is supplied at high reported purity and is commonly used as an intermediate, reagent, or component in assay development.

  • High purity suitable for research applications (reported ~99.8% by COA).
  • Chemical formula C9H13NO; molecular weight 151.21 g/mol.
  • Soluble in DMSO, facilitating stock solution preparation.
  • Useful as an intermediate in organic synthesis and as a reagent in biochemical research.
  • Offered in multiple lab-scale pack sizes, including 50 g.

Catalog No. 50-003-01647 Supplier Medchemexpress LLC Supplier No. HYW01783450G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(Benzyloxy)-1-ethanamine is a small-molecule biochemical reagent used as an organic building block and reagent in life-science and synthetic chemistry research. It is supplied at high reported purity and is commonly used as an intermediate, reagent, or component in assay development.

  • High purity suitable for research applications (reported ~99.8% by COA).
  • Chemical formula C9H13NO; molecular weight 151.21 g/mol.
  • Soluble in DMSO, facilitating stock solution preparation.
  • Useful as an intermediate in organic synthesis and as a reagent in biochemical research.
  • Offered in multiple lab-scale pack sizes, including 50 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.