Learn More
Medchemexpress LLC Ethanaminium, 2-(acetyloxy)-N,N,N-tri(methyl-d3)-, chloride | 344298-95-9 | MFCD01074064 | 99.0% | 190.72 g/mol | C7H7D9ClNO2 | 10 MG
SDP
Catalog No. 5000297401
Encompass_Preferred
Acetylcholine-d9 chloride is the deuterium-labeled form of acetylcholine chloride intended for use as a tracer and internal standard in analytical and biochemical research. It acts as a cholinergic agonist and is suitable for quantitative analysis by NMR, GC-MS, and LC-MS.
  • Deuterium-labeled tracer for quantitative analysis.
  • High purity (99.0%).
  • Molecular weight 190.72 g/mol.
  • White to off-white solid, chloride salt.
  • Suitable for NMR, GC-MS, and LC-MS workflows.
  • Store sealed at 4°C; in solvent store at -80°C for long term.
Catalog No. 50-002-97401 Supplier Medchemexpress LLC Supplier No. HYB0282S110MG
Acetylcholine-d9 chloride is the deuterium-labeled form of acetylcholine chloride intended for use as a tracer and internal standard in analytical and biochemical research. It acts as a cholinergic agonist and is suitable for quantitative analysis by NMR, GC-MS, and LC-MS.
  • Deuterium-labeled tracer for quantitative analysis.
  • High purity (99.0%).
  • Molecular weight 190.72 g/mol.
  • White to off-white solid, chloride salt.
  • Suitable for NMR, GC-MS, and LC-MS workflows.
  • Store sealed at 4°C; in solvent store at -80°C for long term.