Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethanaminium, 2-(acetylthio)-N,N,N-trimethyl-, chloride (1:1) | 6050-81-3 | MFCD00038727 | 98.0% | 197.73 | C7H16ClNOS | 25 G
SDP

Catalog No. 5000283467
Encompass_Preferred

Acetylthiocholine chloride is a biochemical assay substrate commonly used to measure cholinesterase activity in enzymatic and diagnostic assays. It is supplied as a white to off-white solid, characterized by NMR, and accompanied by certificate of analysis information. Store sealed and protected from moisture; when prepared in solvent, follow recommended low-temperature storage for long-term stability.

  • Used as a substrate for cholinesterase activity assays.
  • Purity 98.0% (NMR).
  • CAS number 6050-81-3.
  • Molecular weight 197.73.
  • White to off-white solid appearance.
  • Available in multiple sizes, including 25 g.
  • Store sealed, away from moisture; in solvent, -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-002-83467 Supplier Medchemexpress LLC Supplier No. HYW14095325G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acetylthiocholine chloride is a biochemical assay substrate commonly used to measure cholinesterase activity in enzymatic and diagnostic assays. It is supplied as a white to off-white solid, characterized by NMR, and accompanied by certificate of analysis information. Store sealed and protected from moisture; when prepared in solvent, follow recommended low-temperature storage for long-term stability.

  • Used as a substrate for cholinesterase activity assays.
  • Purity 98.0% (NMR).
  • CAS number 6050-81-3.
  • Molecular weight 197.73.
  • White to off-white solid appearance.
  • Available in multiple sizes, including 25 g.
  • Store sealed, away from moisture; in solvent, -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.