Learn More
Medchemexpress LLC Ethanaminium, 2-hydroxy-N-(iodomethyl)-N,N-dimethyl-, iodide (1:1) | 28508-22-7 | MFCD20489393 | 98.0% | 356.97 | C5H13I2NO | 100 MG
SDP
Catalog No. 5000283656
Encompass_Preferred
Iodocholine iodide is the iodide salt of choline used as a biochemical reagent and non-radioactive reference for radiolabeled choline. It is described as a metabolizable, non-toxic "green" catalyst useful for catalyzing free radical polymerization and is supplied for laboratory assay and research applications.
  • High purity: 97.99%.
  • Molecular formula C5H13I2NO and molecular weight 356.97 g/mol.
  • Suitable as a non-radioactive reference for PET imaging research.
  • Functions as a metabolizable, low-toxicity catalyst for free radical polymerization.
  • Available in multiple pack sizes including 100 mg for laboratory use.
Catalog No. 50-002-83656 Supplier Medchemexpress LLC Supplier No. HYW130177100MG
Iodocholine iodide is the iodide salt of choline used as a biochemical reagent and non-radioactive reference for radiolabeled choline. It is described as a metabolizable, non-toxic "green" catalyst useful for catalyzing free radical polymerization and is supplied for laboratory assay and research applications.
  • High purity: 97.99%.
  • Molecular formula C5H13I2NO and molecular weight 356.97 g/mol.
  • Suitable as a non-radioactive reference for PET imaging research.
  • Functions as a metabolizable, low-toxicity catalyst for free radical polymerization.
  • Available in multiple pack sizes including 100 mg for laboratory use.